Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 11
This gene likely encodes a member of the secernin family of proteins. A similar protein in rat functions in regulation of exocytosis in mast cells. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Secernin-1
Gene Aliases: KIAA0193; SCRN1; SES1
UniProt ID: (Human) Q12765, (Rat) Q6AY84
Entrez Gene ID: (Human) 9805, (Rat) 502776
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support