Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASSSPDSPC SCDCFVSVPP ASAIPAVIFA KNSDRPRDEV QEVVFVPAGT HTPGSRLQCT YIEVEQVSKT HAVILSRPSW LWGAEMGANE HGVCIGNEAV WTREPVGEGE ALLGMDLLRL ALERSSSAQE ALHVITGLLE HYGQGGNCLE DAAPFSYHST FLLADRTEAW VLETAGRLWA AQRIQEGARN ISNQLSIGTD ISAQHPELRT HAQAKGWWDG QGAFDFAQIF SLTQQPVRME AAKARFQAGR ELLRQRQGGI TAEVMMGILR DKESGICMDS GGFRTTASMV SVLPQDPTQP CVHFLTATPD PSRSVFKPFI FGVGVAQAPQ VLSPTFGAQD PVRTLPRFQT QVDRRHTLYR GHQAALGLME RDQDRGQQLQ QKQQDLEQEG LEATQGLLAG EWAPPLWELG GLFQAFVKRE SQAYA
SCRN2 is a member of the secernin family of proteins which includes the homologous SCRN1, a protein that was first identified as being involved in the regulation of exocytosis from peritoneal mast cells. Little is known of SCRN2, but studies have shown that SCRN1 may possess epitopes that function as tumor-associated antigens in gastric cancers and increased SCRN1 expression in patients with colorectal cancer correlated with poor prognosis, suggesting that SCRN2 may also be involved in protein secretion or the regulation of cell growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Secernin-2
Gene Aliases: SCRN2; Ses2
UniProt ID: (Human) Q96FV2, (Rat) Q6AYR8
Entrez Gene ID: (Human) 90507, (Rat) 360612
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support