Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VLSIKQRASF KIKDAKCRLH LRNKGKTEEA GRITGPGGAP CSECQVTFIH LKCDSSRKGK GRRARTPPGK EVTRLTLELE AEVRAEETTA S
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
SCUBE3 is a member of a family of secreted glycoproteins that contain N-terminal EGF-like repeats and C-terminal cysteine-rich motifs and CUB domain and is highly expressed in primary osteoblasts and bones, and to a lesser extent in heart. Other studies have shown that overexpression of SCUBE3 in mice induced cardiac hypertrophy, suggesting that it may also play a role in the regulation of cardiac growth. SCUBE3 has been shown to be an endogenous TGF-Beta receptor ligand and is thought to promote lung cancer cell mobility and invasiveness. In lung cancer cells, the secreted SCUBE3 protein was cleaved by MMP2 and MMP9, allowing the activation of the TGF-Beta receptor, the increase of Smad2/3 transcriptional activity and the upregulation of expression of proteins such as TGF-Beta1, VEGF, Snail, and Slug.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CUB domain and EGF-like repeat containing 3; signal peptide; Signal peptide, CUB and EGF-like domain-containing protein 3
Gene Aliases: CEGF3; SCUBE3
UniProt ID: (Human) Q8IX30
Entrez Gene ID: (Human) 222663
Molecular Function:
extracellular matrix glycoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support