Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a major catalytic subunit of succinate-ubiquinone oxidoreductase, a complex of the mitochondrial respiratory chain. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. Mutations in this gene have been associated with a form of mitochondrial respiratory chain deficiency known as Leigh Syndrome. A pseudogene has been identified on chromosome 3q29.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Flavoprotein subunit of complex II; Fp; succinate dehydrogenase [ubiquinone] flavoprotein subunit; Succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial; succinate dehydrogenase complex subunit A, flavoprotein (Fp); succinate dehydrogenase complex, subunit A, flavoprotein (Fp)
Gene Aliases: 1500032O14Rik; 2310034D06Rik; 4921513A11; C81073; CMD1GG; FP; PGL5; SDH1; SDH2; SDHA; SDHF
UniProt ID: (Human) P31040, (Rat) Q920L2, (Mouse) Q8K2B3
Entrez Gene ID: (Human) 6389, (Rat) 157074, (Mouse) 66945
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support