Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAALLLRHVG RHCLRAHFSP QLCIRNAVPL GTTAKEEMER FWNKNIGSNR PLSPHITIYS WSLPMAMSIC HRGTGIALSA GVSLFGMSAL LLPGNFESYL ELVKSLCLGP ALIHTAKFAL VFPLMYHTWN GIRHLMWDLG KGLKIPQLYQ SGVVVLVLTV LSSMGLAAM
This gene encodes one of four nuclear-encoded subunits that comprise succinate dehydrogenase, also known as mitochondrial complex II, a key enzyme complex of the tricarboxylic acid cycle and aerobic respiratory chains of mitochondria. The encoded protein is one of two integral membrane proteins that anchor other subunits of the complex, which form the catalytic core, to the inner mitochondrial membrane. There are several related pseudogenes for this gene on different chromosomes. Mutations in this gene have been associated with paragangliomas. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYBL; cytochrome B large subunit of complex II; Integral membrane protein CII-3; integral membrane protein CII-3b; large subunit of cytochrome b; QPs-1; QPs1; succinate dehydrgenase cytochrome b; succinate dehydrogenase 3, integral membrane subunit; Succinate dehydrogenase complex subunit C; succinate dehydrogenase complex subunit C integral membrane protein 15kDa; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kD; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa; Succinate dehydrogenase cytochrome b560 subunit, mitochondrial; Succinate-ubiquinone oxidoreductase cytochrome B large subunit
Gene Aliases: CYB560; CYBL; PGL3; QPS1; SDH3; SDHC
UniProt ID: (Human) Q99643
Entrez Gene ID: (Human) 6391
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support