Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
SDSL has low serine dehydratase and threonine dehydratase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH 2; Serine dehydratase 2; serine dehydratase related sequence 1; Serine dehydratase-like; TDH
Gene Aliases: cSDH; SDH 2; SDS-RS1; SDSL; TDH
UniProt ID: (Human) Q96GA7
Entrez Gene ID: (Human) 113675
Molecular Function:
dehydratase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support