Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in mouse and shares 95% identity with rat sequence.
Specificity of this antibody: SEC22C.
May be involved in vesicle transport between the ER and the Golgi complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SEC22 vesicle trafficking protein homolog C; SEC22 vesicle trafficking protein-like 3; SEC22 vesicle trafficking protein-like C; SEC22 vesicle trafficking protein-like protein C; SEC22 vesicle-trafficking protein homolog C; SEC22 vesicle-trafficking protein-like 3; secretion deficient 22C; Vesicle-trafficking protein SEC22c
Gene Aliases: 4932412K21; 5930407I15Rik; C530046H07; SEC22C; SEC22L3; UNQ459/PRO784
UniProt ID: (Human) Q9BRL7, (Mouse) Q8BXT9
Entrez Gene ID: (Human) 9117, (Mouse) 215474
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support