Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: SEC31b.
This gene encodes a protein of unknown function. The protein has moderate similarity to rat VAP1 protein which is an endosomal membrane-associated protein, containing a putative Ca2+/calmodulin-dependent kinase II phosphorylation site.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein transport protein Sec31B; sc31b; SEC31-like 2; SEC31-like protein 2; SEC31-related protein B
Gene Aliases: Gm341; Sec31b; Sec31l2
UniProt ID: (Mouse) Q3TZ89
Entrez Gene ID: (Mouse) 240667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support