Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Has chemorepulsive activities for sympathetic axons. Ligand of NRP2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Semaphorin sem2; Semaphorin-3G
Gene Aliases: sem2; SEMA3G
UniProt ID: (Human) Q9NS98
Entrez Gene ID: (Human) 56920
Molecular Function:
membrane-bound signaling molecule
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support