Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPRAPHFMPL LLLLLLLSLP HTQAAFPQDP LPLLISDLQG TSPLSWFRGL EDDAVAAELG LDFQRFLTLN RTLLVAARDH VFSFDLQAEE EGEGLVPNKY LTWRSQDVEN CAVRGKLTDE CYNYIRVLVP WDSQTLLACG TNSFSPVCRS YGITSLQQEG EELSGQARCP FDATQSNVAI FAEGSLYSAT AADFQASDAV VYRSLGPQPP LRSAKYDSKW LREPHFVQAL EHGDHVYFFF REVSVEDARL GRVQFSRVAR VCKRDMGGSP RALDRHWTSF LKLRLNCSVP GDSTFYFDVL QALTGPVNLH GRSALFGVFT TQTNSIPGSA VCAFYLDEIE RGFEGKFKEQ RSLDGAWTPV SEDRVPSPRP GSCAGVGGAA LFSSSRDLPD DVLTFIKAHP LLDPAVPPVT HQPLLTLTSR ALLTQVAVDG MAGPHSNITV MFLGSNDGTV LKVLTPGGRS GGPEPILLEE IDAYSPARCS GKRTAQTARR IIGLELDTEG HRLFVAFSGC IVYLPLSRCA RHGACQRSCL ASQDPYCGWH SSRGCVDIRG SGGTDVDQAG NQESMEHGDC QDGATGSQSG PGDSAYGVRR DLPPASASRS VPIPLLLASV AAAFALGASV SGLLVSCACR RAHRRRGKDI ETPGLPRPLS LRSLARLHGG GPEPPPPSKD GDAVQTPQLY TTFLPPPEGV PPPELACLPT PESTPELPVK HLRAAGDPWE WNQNRNNAKE GPGRSRGGHA AGGPAPRVLV RPPPPGCPGQ AVEVTTLEEL LRYLHGPQPP RKGAEPPAPL TSRALPPEPA PALLGGPSPR PHECASPLRL DVPPEGRCAS APARPALSAP APRLGVGGGR RLPFSGHRAP PALLTRVPSG GPSRYSGGPG KHLLYLGRPE GYRGRALKRV DVEKPQLSLK PPLVGPSSRQ AVPNGGRFNF
This gene product is a member of the semaphorin family of proteins. Semaphorins represent important molecular signals controlling multiple aspects of the cellular response that follows CNS injury, and thus may play an important role in neural regeneration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C; Sema Y; Semaphorin-6C; Semaphorin-Y
Gene Aliases: KIAA1869; m-SemaY; m-SemaY2; SEMA6C; SEMAY
UniProt ID: (Human) Q9H3T2
Entrez Gene ID: (Human) 10500
Molecular Function: membrane-bound signaling molecule intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support