Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: Mouse brain, K562, SHSY-5Y, HUVEC, HepG2, human colon cancer tissue, human kidney tissue, rat brain tissue.
Septin 2 is a filament-forming cytoskeletal GTPase. Septin 2 is required for normal organization of the actin cytoskeleton and plays a role in the biogenesis of polarized columnar-shaped epithelium by maintaining polyglutamylated microtubules, thus facilitating efficient vesicle transport, and by impeding MAP4 binding to tubulin. It is required for the progression through mitosis, and it forms a scaffold at the midplane of the mitotic splindle required to maintain CENPE localization at kinetochores and consequently chromosome congression. During anaphase, Septin 2 may be required for chromosome segregation and spindle elongation. Septin 2 also plays a role in ciliogenesis and collective cell movements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-Sep; NEDD-5; Neural precursor cell expressed developmentally down-regulated protein 5; neural precursor cell expressed, developmentally down-regulated 5; Septin-2
Gene Aliases: DIFF6; hNedd5; KIAA0158; NEDD-5; NEDD5; Pnutl3; SEPT2; SEPTIN2
UniProt ID: (Human) Q15019
Entrez Gene ID: (Human) 4735
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support