Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSFCSFFGG EVFQNHFEPG VYVCAKCGYE LFSSRSKYAH SSPWPAFTET IHADSVAKRP EHNRSEALKV SCGKCGNGLG HEFLNDGPKP GQSRFUIFSS SLKFVPKGKE TSASQG (1-115 aa encoded by B C003127)
This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This protein belongs to the methionine sulfoxide reductase B (MsrB) family, and it is expressed in a variety of adult and fetal tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: methionine sulfoxide red. domain; methionine sulfoxide reductase b; Methionine-R-sulfoxide reductase B1; MGC3344; MsrB1; Selenoprotein R; Selenoprotein X; selenoprotein X 1; selenoprotein X, 1; SelR; SelX
Gene Aliases: D17Wsu82e; HSPC270; MSRB1; SELR; SELX; SepR; SEPX1
UniProt ID: (Human) Q9NZV6, (Rat) Q52KJ8, (Mouse) Q9JLC3
Entrez Gene ID: (Human) 51734, (Rat) 685059, (Mouse) 27361
Molecular Function:
oxidoreductase
reductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support