Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSLAAYCVI CCRRIGTSTS PPKSGTHWRD IRNIIKFTGS LILGGSLFLT YEVLALKKAV TLDTQVVERE KMKSYIYVHT VSLDKGENHG IAWQARKELH KAVRKVLATS AKILRNPFAD PFSTVDIEDH ECAVWLLLRK SKSDDKTTRL EAVREMSETH HWH (1-162 aa encoded by BC001705)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein SERAC1; Serine active site-containing protein 1; srac1
Gene Aliases: 4930511N22Rik; D17Ertd141e; SERAC1
UniProt ID: (Human) Q96JX3, (Mouse) Q3U213
Entrez Gene ID: (Human) 84947, (Mouse) 321007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support