Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3'-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromodomain helicase DNA binding protein 3 interacting protein; PAI RBP 1; PAI-1 mRNA binding protein; PAI-RBP1; PAI1 RNA-binding protein 1; plasminogen activator inhibitor 1 RNA binding protein; Plasminogen activator inhibitor 1 RNA-binding protein; SERPINE1 mRNA-binding protein 1
Gene Aliases: CGI-55; CHD3IP; HABP4L; PAI-RBP1; PAIRBP1; SERBP1
UniProt ID: (Human) Q8NC51
Entrez Gene ID: (Human) 26135
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support