Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blots, no cross-reactivity with recombinant human (rh) Serpin A1, A3, A4, A5, A6, B8, 9, C1, D1, E1, F2, G1, I1, I2, recombinant mouse (rm) Serpin E2, F1, recombinant rat Serpin B6, rhA1 Antitrypsin, rhAngiotensinogin, or rmPEDF is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
Vaspin is a newly described adipocytokine expressed predominantly in visceral white adipose tissues. Structure analysis of vaspin predicts the presence of three beta-sheets, nine alpha-helices, and one central loop, which are distinctive structural features of Serpin family members. The serpins are irreversible ("suicidal") serine-protease inhibitors, characterized by having more than 30% sequence homology with alpha1-antitrypsin and a conserved tertiary structure, which contains an exposed reactive center loop that acts as a pseudo-substrate for the target proteinase. Members of this family play an important role in a number of fundamental biological processes, including blood coagulation, fibrinolysis, complement activation, angiogenesis, inflammation, and tumor suppression. In humans, the serpins represent approximately 2% of total serum proteins, of which 70% is alpha1-antitrypsin. Vaspin exhibits 40.2% sequence identity with alpha1-antitrypsin. Yet, its protease inhibitory activity is still unknown. Vaspin mRNA expression in visceral fat is positively correlated with BMI and percent of body fat. Administration of vaspin to obese mice improved glucose tolerance and insulin sensitivity, reflected by normalized blood glucose levels. It also led to the reversal of altered expression of diabetes-relevant adipocytokines, including leptin, adiponectin, resistin, and TNF-alpha. These findings suggest a potential clinical use for Vaspin in ameliorating certain aberrations seen in the diabetic/obesity metabolic syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a12; OL-64; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12; Serpin A12; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12; Vaspin; Visceral adipose tissue-derived serine protease inhibitor; Visceral adipose-specific serpin
Gene Aliases: OL-64; SERPINA12
UniProt ID: (Human) Q8IW75
Entrez Gene ID: (Human) 145264
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support