Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Antibody detects endogenous levels of total SESN2.
This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hypoxia induced gene 95; hypoxia-induced; Hypoxia-induced gene; Sestrin-2
Gene Aliases: HI95; RGD1566319; SES2; SESN2; SEST2
UniProt ID: (Human) P58004, (Mouse) P58043
Entrez Gene ID: (Human) 83667, (Rat) 502988, (Mouse) 230784
Molecular Function: peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support