Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HYHALASFV FGSGINPERD PEISNGFRLI SVNNFCVCDL ANDNNIENAS LSGSNFGIVD SLSEL (199-262 aa encoded by BC017296)
May function as an intracellular leucine sensor that negatively regulates the TORC1 signaling pathway (PubMed:25263562). May also regulate the insulin-receptor signaling pathway through activation of TORC2. This metabolic regulator may also play a role in protection against oxidative and genotoxic stresses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sestrin-3
Gene Aliases: SESN3; SEST3
UniProt ID: (Human) P58005
Entrez Gene ID: (Human) 143686
Molecular Function:
peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support