Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QNRPDSRLGK TELSFSSSCE IPHVDGLHSS EELRNLGWDF SQEKPSTTYQ QPDSSYGACG GHKYQQNAEQ YGGTRDYWQG NGYWDPRSGR PPGTGVVYDR TQGQVPDSLT DDREEEENWD QQDGSHFSDQ SDKFLLSLQK DKGSVQAPEI SSNSIKDTLA VNEKKDFSKN LEKNDIKDRG PLKKRRQEIE SDSESDGELQ DRKKVRVE
Positive Samples: Mouse brain, Rat brain; Cellular Location: Chromosome, Nucleus
Histone methyltransferase that specifically trimethylates 'Lys-36' of histone H3 (H3K36me3) using dimethylated 'Lys-36' (H3K36me2) as substrate. Represents the main enzyme generating H3K36me3, a specific tag for epigenetic transcriptional activation. Plays a role in chromatin structure modulation during elongation by coordinating recruitment of the FACT complex and by interacting with hyperphosphorylated POLR2A. Acts as a key regulator of DNA mismatch repair in G1 and early S phase by generating H3K36me3, a mark required to recruit MSH6 subunit of the MutS alpha complex: early recruitment of the MutS alpha complex to chromatin to be replicated allows a quick identification of mismatch DNA to initiate the mismatch repair reaction. H3K36me3 also plays an essential role in the maintenance of a heterochromatic state, by recruiting DNA methyltransferase DNMT3A. H3K36me3 is also enhanced in intron-containing genes, suggesting that SETD2 recruitment is enhanced by splicing and that splicing is coupled to recruitment of elongating RNA polymerase. Required during angiogenesis. Recruited to the promoters of adenovirus 12 E1A gene in case of infection, possibly leading to regulate its expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16420; FLJ22472; FLJ23184; FLJ45883; Histone-lysine N-methyltransferase SETD2; kinesin family member 9; Lysine N-methyltransferase 3A; Protein-lysine N-methyltransferase SETD2; SET domain-containing protein 2
Gene Aliases: 4921524K10Rik; BC031601; Kiaa1732; Kif9; KMT3A; Setd2
UniProt ID: (Mouse) E9Q5F9
Entrez Gene ID: (Rat) 316013, (Mouse) 235626
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support