Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Control: K562, THP-1, HL-60
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SETD6 (SET domain-containing protein 6) is a 473 amino acid protein that belongs to the SETD6 family. Containing one SET domain, SETD6 is structurally similar to the Rubisco large subunit methyltransferase. SETD6 is a proteinlysine N-methyltransferase that specifically monomethylates "Lys-310" of the NFκB p65 subunit of NFκB complex, leading to down-regulate NFκB transcription factor activity. SETD6-mediated methylation renders NFκB p65 inert and attenuates NFκB p65-driven transcriptional programs, including inflammatory responses in primary immune cells. The SETD6-initiated lysine-methylation signaling cascade acts to restrain activation of NFκB-mediated inflammatory responses in diverse cell types.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-lysine methyltransferase SETD6; SET domain-containing protein 6
Gene Aliases: SETD6
UniProt ID: (Human) Q8TBK2
Entrez Gene ID: (Human) 79918
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support