Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSLPGCIG LDAATATVES EEIAELQQAV VEELGISMEE LRHFIDEELE KMDCVQQRKK QLAELETWVI QKESEVAHVD QLFDDASRAV TNCESLVKDF YSKLGLQYRD SSSEDESSRP TEIIEIPDED DDVLSIDSGD AGSRTPKDQK LREAMAALRK SAQDVQKFMD AVNKKSSSQD LHKGTLSQMS GELSKDGDLI VSMRILGKKR TKTWHKGTLI AIQTVGPGKK YKVKFDNKGK SLLSGNHIAY DYHPPADKLY VGSRVVAKYK DGNQVWLYAG IVAETPNVKN KLRFLIFFDD GYASYVTQSE LYPICRPLKK TWEDIEDISC RDFIEEYVTA YPNRPMVLLK S (1-350 aa encoded by BC009362)
This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N(6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERG-associated protein with a SET domain, ESET; ERG-associated protein with SET domain; ESET; H3-K9-HMTase 4; Histone H3-K9 methyltransferase 4; Histone-lysine N-methyltransferase SETDB1; histone-lysine N-methyltransferase, H3lysine-9 specific 4; Lysine N-methyltransferase 1E; SET domain bifurcated 1; SET domain ERG-associated histone methyltransferase; SET domain, bifurcated 1; tudor domain containing 21
Gene Aliases: AU022152; ESET; H3-K9-HMTase4; KG1T; KIAA0067; KMT1E; mKIAA0067; SETDB1; TDRD21
UniProt ID: (Human) Q15047
Entrez Gene ID: (Human) 9869, (Mouse) 84505, (Rat) 689883
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support