Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
SEZ6L (seizure related 6 homolog-like) is a 1,024 amino acid single-pass type I membrane protein that localizes to the endoplasmic reticulum. Widely expressed, SEZ6L is a candidate tumor suppressor gene and may contribute to specialized endoplasmic reticulum functions in neurons. SEZ6L contains three CUB domains and five sushi (CCP/SCR) domains, which may be involved in protein-protein interactions and signal transduction. The gene encoding SEZ6L is located on human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, Neurofibromatosis type 2, autism and schizophrenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acupuncture induced gene 1; Acupuncture-induced protein 1-L; Brain-specific receptor-like protein B; BSRP-B; Seizure 6-like protein; seizure related 6 homolog (mouse)-like; seizure related gene 6-like
Gene Aliases: Acig1; AI843918; AIG1; Aig1l; AW539724; BSRP-B; KIAA0927; mKIAA0927; RGD1563628; SEZ6L; UNQ2542/PRO6094
UniProt ID: (Human) Q9BYH1, (Mouse) Q6P1D5
Entrez Gene ID: (Human) 23544, (Mouse) 56747, (Rat) 304554
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support