Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAGPISER NQDATVYVGG LDEKVSEPLL WELFLQAGPV VNTHMPKDRV TGQHQGYGFV EFLSEEDADY AIKIMNMIKL YGKPIRVNKA SAHNKNLDVG ANIFIGNLDP EIDEKLLYDT FSAFGVILQT PKIMRDPDTG NSKGYAFINF ASFDASDAAI EAMNGQYLCN RPITVSYAFK KDSKGERHGS AAERLLAAQN P (1-200 aa encoded by BC004273)
This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC10828; Pre-mRNA-splicing factor SF3b 49 kDa subunit; SAP 49; SAP49MGC10828; SF3b50; spliceosomal protein; spliceosome-associated protein (U2 snRNP); Spliceosome-associated protein 49; Splicing factor 3B subunit 4; splicing factor 3b, subunit 4, 49kD; splicing factor 3b, subunit 4, 49kDa
Gene Aliases: AFD1; Hsh49; SAP49; SF3B4; SF3b49
UniProt ID: (Human) Q15427
Entrez Gene ID: (Human) 10262
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support