Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Secreted frizzled-related protein 1 (SFRP1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: frizzled-related protein; FRP-1; SARP-2; Secreted apoptosis-related protein 2; Secreted frizzled-related protein 1; secreted frizzled-related sequence protein 1; sFRP-1
Gene Aliases: 2210415K03Rik; AW011917; AW107218; AW742929; FRP; FRP-1; FRP1; FrzA; SARP2; sFRP-1; SFRP1
UniProt ID: (Human) O14779, (Mouse) Q8C4U3
Entrez Gene ID: (Human) 6422, (Mouse) 20377, (Rat) 84402
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support