Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total SFRP2.
This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. Methylation of this gene is a potential marker for the presence of colorectal cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FRP 2; FRP-2; MGC128911; MGC153618; MGC53344; Protein SDF5; SARP 1; SARP-1; SDF 5; secreted apoptosis related protein 1; Secreted apoptosis-related protein 1; Secreted frizzled-related protein 2; secreted frizzled-related sequence protein 2; secreted frizzled-related sequence protein 5; sFRP 2; sFRP-2; Sfrp2 protein; stromal cell derived factor 5; testicular tissue protein Li 170
Gene Aliases: AI851596; FKSG12; FRP-2; FRP2; SARP1; SDF-5; Sdf5; SFRP2; UNQ361/PRO697
UniProt ID: (Human) Q96HF1, (Mouse) P97299
Entrez Gene ID: (Human) 6423, (Rat) 310552, (Mouse) 20319
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support