Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: LGVFGLSLYT TERDLREVFS KYGPIADVSI VYDQQSRRSR GFAFVYFENV DDAKEAKERA NGMELDGRRI RVDFSITKRP
SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein phosphatase 1, regulatory subunit 156; Splicing factor, arginine/serine-rich 10; splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila); TRA-2 beta; Transformer-2 protein homolog B; Transformer-2 protein homolog beta
Gene Aliases: Htra2-beta; PPP1R156; SFRS10; SRFS10; TRA2-BETA; TRA2B; TRAN2B
UniProt ID: (Human) D3DNU3
Entrez Gene ID: (Human) 6434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support