Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Samples: NCI-H460, NCI-H125, NIH/3T3, 3T3L1, Mouse intestine; Cellular Location: extracellular matrix, extracellular space, surface film
Immunogen sequence: RGALSLQGSI MTVGEKVFSS NGQSITFDAI QEACARAGGR IAVPRNPEEN EAIASFVKKY NTYAYVGLTE GPSPGDFRYS DGTPVNYTNW YRGEPAGRGK
Surfactant protein A is an innate immune system collectin. The gene encoding SP-A map to mouse chromosome 14. Surfactant protein-A, which plays a role in innate host defense in the lung, is also expressed in the Eustachian tube. We report that the frequency of specific surfactant protein-A haplotypes and genotypes differs between children with recurrent otitis media compared with a control population.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein; Collectin 4; Collectin-4; FLJ50593; FLJ51913; FLJ61144; FLJ77898; FLJ79095; FLJ99559; MGC133365; MGC198590; OTTHUMP00000019928; OTTHUMP00000019929; OTTHUMP00000019930; OTTHUMP00000019931; PSAP; PSP A; PSP-A; Pulmonary surfactant-associated protein A; Pulmonary surfactant-associated protein A1; SP A; SP A1; SP-A1 beta; SP-A1 delta; SP-A1 epsilon; SP-A1 gamma; Surfactant; surfactant associated protein A; surfactant protein A1 variant AB'D' 6A; surfactant protein A1 variant AB'D' 6A2; surfactant protein A1 variant AB'D' 6A3; surfactant protein A1 variant AB'D' 6A4; surfactant protein A1 variant ACD' 6A; surfactant protein A1 variant ACD' 6A2; surfactant protein A1 variant ACD' 6A3; surfactant protein A1 variant ACD' 6A4; surfactant protein A1 variant AD' 6A; surfactant protein A1 variant AD' 6A2; surfactant protein A1 variant AD' 6A3; surfactant protein A1 variant AD' 6A4; surfactant protein A1B; surfactant pulmonary associated protein A1; surfactant, pulmonary-associated protein A1; surfactant, pulmonary-associated protein A1A; surfactant, pulmonary-associated protein A1B; Surfactant-associated protein 1 (pulmonary surfactant protein SP-A); Surfactant-associated protein 1 (pulmonary surfactant protein, SP-A)
Gene Aliases: COLEC4; PSAP; PSP-A; PSPA; Sftp-1; SFTP1; SFTPA; SFTPA1; SFTPA1B; Sftpl; SP-A; SP-A1; SPA; SPA1
UniProt ID: (Human) Q8IWL2, (Rat) P08427
Entrez Gene ID: (Human) 653509, (Rat) 24773, (Mouse) 20387
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support