Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: MSGELPPNIN IKEPRWDQST FIGRANHFFT VTDPRNILLT NEQLESARKI
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 92%; Zebrafish: 85%
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sideroflexin-1; tricarboxylate carrier protein
Gene Aliases: SFXN1; TCC
UniProt ID: (Human) Q9H9B4
Entrez Gene ID: (Human) 94081
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support