Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 35 kDa dystrophin-associated glycoprotein; 35DAG; CMD1LSG-delta; Delta-sarcoglycan; Delta-SG; dystrophin associated glycoprotein, delta sarcoglycan; LGMD2F; MGC22567; placental delta sarcoglycan; sarcoglycan, delta (35kDa dystrophin-associated glycoprotein); sarcoglycan-delta; sarcoglycandelta; SGdelta
Gene Aliases: 35DAG; CMD1L; DAGD; SG-delta; SGCD; SGCDP; SGD
UniProt ID: (Human) Q92629
Entrez Gene ID: (Human) 6444
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support