Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DLRLESPTRS LSMDAPRGVH IQAHAGKIEA LSQMDILFHS SDGMLVLDAE TVCLPKLVQG TWGPSGSSQS LYEICVCPDG KLYLSVAGVS TTCQEHSHIC
Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 35 kDa dystrophin-associated glycoprotein; 35DAG; 35kD dystrophin-associated glycoprotein; Gamma-sarcoglycan; Gamma-SG; sarcoglycan, gamma (35kDa dystrophin-associated glycoprotein); TYPE
Gene Aliases: 35DAG; A4; DAGA4; DMDA; DMDA1; gamma-SG; LGMD2C; MAM; SCARMD2; SCG3; SGCG
UniProt ID: (Human) Q13326
Entrez Gene ID: (Human) 6445
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support