Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 35 kDa dystrophin-associated glycoprotein; 35DAG; 35kD dystrophin-associated glycoprotein; Gamma-sarcoglycan; Gamma-SG; sarcoglycan, gamma (35kD dystrophin-associated glycoprotein); sarcoglycan, gamma (35kDa dystrophin-associated glycoprotein); TYPE
Gene Aliases: 35DAG; 35kDa; 5430420E18Rik; A4; AI642964; DAGA4; DMDA; DMDA1; gamma-SG; LGMD2C; MAM; SCARMD2; SCG3; SGCG
UniProt ID: (Human) Q13326, (Mouse) P82348
Entrez Gene ID: (Human) 6445, (Mouse) 24053
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support