Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MTVKTEAAKG TLTYSRMRGM VAILIAFMKQ RRMGLNDFIQ KIANNSYACK HPEVQSILKI SQPQEPELMN ANPSPPPSPS QQINLGPSSN PHAKPSDFHF LKVIGKGSFG KVLLARHKAE EVFYAVKVLQ KKAILKKKEE KHIMSERNVL LKNVKHPFLV GLHFSFQTAD KLYFVLDYIN GGELFYHLQR ERCFLEPRAR FYAAEIASAL GYLHSLNIVY RDLKPENILL DSQGHIVLTD FGLCKENIEH NSTTSTFCGT PEYLAPEVLH KQPYDRTVDW WCLGAVLYEM LYGLPPFYSR NTAEMYDNIL NKPLQLKPNI TNSARHLLEG LLQKDRTKRL GAKDDFMEIK SHVFFSLINW DDLINKKITP PFNPNVSGPN DLRHFDPEFT EEPVPNSIGK SPDSVLVTAS VKEAAEAFLG FSYAPPTDSF L
SGK (SGK1) is a serine/threonine kinase that may be a transcriptional target for p38/MAPK in hepatoma cells. SGK plays an important role in cellular stress response and cell survival. SGK1 is transcriptionally activated by numerous stimuli, including serum, glucocorticoids, osmotic stress, and hormones. It phosphorylates and negatively regulates pro-apoptotic FOXO3A. SGK1 is thought to play an essential function in nephropathy and cardiovascular development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: serine/threonine protein kinase SGK; Serine/threonine-protein kinase Sgk1; Serum/glucocorticoid-regulated kinase 1; Sgk1 variant i3
Gene Aliases: SGK; SGK1
UniProt ID: (Human) O00141
Entrez Gene ID: (Human) 6446
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support