Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SPINWDDLYH KRLTPPFNPN VTGPADLKHF DPEFTQEAVS KSIGCTPDTV ASSSGASSAF LGFSYAPEDD DILDC
SGK2 is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGK2 is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. Although little is known about the transcriptional regulation of SGK2, the protein has been shown to be phosphorylated and activated by 3 phosphoinositide dependent protein kinase 1 (PDK1), and to a lesser extent insulin like growth factor 1, in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine/threonine-protein kinase Sgk2; serum/glucocorticoid regulated kinase 2; Serum/glucocorticoid-regulated kinase 2
Gene Aliases: AI098171; AW146006; dJ138B7.2; H-SGK2; SGK2; Sgkl
UniProt ID: (Human) Q9HBY8, (Mouse) Q9QZS5
Entrez Gene ID: (Human) 10110, (Mouse) 27219
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support