Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: DNGIPFPSGR TNLYWPGTAE PLLVSSPEHP KRWGQVSEAY VSLLDLTPTI LDWFSIPYPS YAIFGSKTIH LTGRSLLPAL EAEPLWATVF GSQSHHEVTM SYPMRSVQHR HFRLVHNLNF KMPFPIDQDF YVSPTFQDLL NRTTAGQPTG WYKDLRHYYY RARWELYDRS RDPHETQNLA TDPRFAQLLE MLRDQLAKWQ WETHDPWVCA PDGVLEEKLS PQCQPLHNEL; Positive Samples: MCF7, 22Rv1, HepG2, Mouse liver, Mouse testis; Cellular Location: Lysosome
SGSH is one of several enzymes involved in the lysosomal degradation of heparan sulfate. Mutations in this gene are associated with Sanfilippo syndrome A, one type of the lysosomal storage disease mucopolysaccaridosis III, which results from impaired degradation of heparan sulfate. Transcripts of varying sizes have been reported but their biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heparan sulfate sulfatase; mucopolysaccharidosis type IIIA; N-sulfoglucosamine sulfohydrolase; N-sulphoglucosamine sulphohydrolase; Sulfoglucosamine sulfamidase; Sulphamidase
Gene Aliases: 4632406A19Rik; HSS; MPS3A; SFMD; SGSH
UniProt ID: (Human) P51688
Entrez Gene ID: (Human) 6448, (Rat) 688293, (Mouse) 27029
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support