Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDNKKRLAY AIIQFLHDQL RHGGLSSDAQ ESLEVAIQCL ETAFGVTVED SDLALPQTLP EIFEAAATGK EMPQDLRSPA RTPPSEEDSA EAERLKTEGN EQMKVENFEA AVHFYGKAIE LNPANAVYFC NRAAAYSKLG NYAGAVQDCE RAICIDPAYS KAYGRMGLAL SSLNKHVEAV AYYKKALELD PDNETYKSNL KIAELKLREA PSPTGGVGSF DIAGLLNNPG FMSMASNLMN NPQIQQLMSG MISGGNNPLG TPGTSPSQND LASLIQAGQQ FAQQMQQQNP ELIEQLRSQI RSRTPSASND DQQE (1-313 aa encoded by BC008885)
This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha h UBP; Alpha-SGT; protein containing three tetratricopeptide repeats; Small glutamine-rich protein with tetratricopeptide repeats 1; small glutamine-rich tetratricopeptide repeat (TPR) containing protein; small glutamine-rich tetratricopeptide repeat (TPR) containing protein (SGT); small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha; Small glutamine-rich tetratricopeptide repeat-containing protein alpha; UBP; Vpu-binding protein
Gene Aliases: 5330427H01Rik; AI194281; alphaSGT; D10Ertd190e; hSGT; SGT; SGT1; SGTA; Stg
UniProt ID: (Human) O43765, (Mouse) Q8BJU0, (Rat) O70593
Entrez Gene ID: (Human) 6449, (Mouse) 52551, (Rat) 64667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support