Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDLPYYHGRL TKQDCETLLL KEGVDGNFLL RDSESIPGVL CLCVSFKNIV YTYRIFREKH GYYRIQTAEG SPKQVFPSLK ELISKFEKPN QGMVVHLLKP IKRTSPSLRW RGLKLELETF VNSNSDYVDV LP
SH2D1B plays a role in controlling signal transduction through at least four receptors, CD84, CD150, CD229 and CD244, expressed on the surface of professional antigen-presenting cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAT-2; EWS/FLI1-activated transcript 2; SH2 domain-containing molecule EAT2; SH2 domain-containing protein 1B
Gene Aliases: EAT2; SH2D1B
UniProt ID: (Human) O14796
Entrez Gene ID: (Human) 117157
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support