Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KKANELLLST GMPGSFLIRV SERIKGYALS YLSEDGCKHF LIDASADAYS FLGVDQLQHA TLADLVEYHK EEPITSLGKE LLLYPCGQQD QLPDYLELFE
SH2D4A inhibits estrogen-induced cell proliferation by competing with PLCG for binding to ESR1, blocking the effect of estrogen on PLCG and repressing estrogen-induced proliferation. SH2D4A may play a role in T-cell development and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20967; Protein phosphatase 1 regulatory subunit 38; protein phosphatase 1, regulatory subunit 38; Protein SH(2)A; SH2 domain-containing protein 4A
Gene Aliases: PPP1R38; SH2A; SH2D4A
UniProt ID: (Human) Q9H788
Entrez Gene ID: (Human) 63898
Molecular Function: defense/immunity protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support