Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
SH3BGR is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 21-GARP; 21-glutamic acid-rich protein; SH3 domain binding glutamate-rich protein; SH3 domain binding glutamic acid-rich protein; SH3 domain-binding glutamic acid-rich protein; SH3-binding domain and glutamic acid-rich protein; SH3BGR protein
Gene Aliases: 21-GARP; SH3BGR
UniProt ID: (Human) D3DSI2
Entrez Gene ID: (Human) 6450
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support