Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SH3BGRL3 could act as a modulator of glutaredoxin biological activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 297; SH3 domain binding glutamate-rich protein like 3; SH3 domain binding glutamic acid-rich protein like 3; SH3 domain-binding glutamic acid-rich-like protein 3; SH3 domain-binding protein 1; SH3BGRL3-like protein; SH3BP-1; TIP-B1; TNF inhibitory protein; TNF inhibitory protein B1
Gene Aliases: HEL-S-297; P1725; SH3BGRL3; SH3BP-1; TIP-B1
UniProt ID: (Human) Q9H299
Entrez Gene ID: (Human) 83442
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support