Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 13
Binds differentially to the SH3 domains of certain proteins of signal transduction pathways. This protein binds preferentially to the ABL1 proto-oncogene, SRC and GRB2. Shows GAP activity for Rac-related proteins but not for Rho- or Ras-related proteins. It inhibits PDGF-induced membrane ruffling mediated by Rac.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3BP-1; OTTHUMP00000028981; SH3 domain-binding protein 1; SH3-domain binding protein 1
Gene Aliases: ARHGAP43; SH3BP1
UniProt ID: (Human) Q9Y3L3
Entrez Gene ID: (Human) 23616
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support