Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LPNSVFVNTT ESCEVERLFK ATSPRGEPQD GLYCIRNSST KSGKVLVVWD ETSNKVRNYR IFEKDSKFYL EGEVLFVSVG SMVEHY
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.
The protein encoded by this gene has an N-terminal pleckstrin homology (PH) domain, an SH3-binding proline-rich region, and a C-terminal SH2 domain. The protein binds to the SH3 domains of several proteins including the ABL1 and SYK protein tyrosine kinases, and functions as a cytoplasmic adaptor protein to positively regulate transcriptional activity in T, natural killer (NK), and basophilic cells. Mutations in this gene result in cherubism. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3BP-2; Abl-SH3 binding protein 2; FLJ42079; SH3 domain-binding protein 2; SH3-domain binding protein 2; TNFAIP3 interacting protein 2
Gene Aliases: 3BP-2; 3BP2; CRBM; CRPM; RES4-23; SH3BP2
UniProt ID: (Human) P78314
Entrez Gene ID: (Human) 6452
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support