Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Ovarian cancer
Cellular location: Mitochondria.
Inhibits the auto- and transphosphorylation activity of BTK. Plays a negative regulatory role in BTK-related cytoplasmic signaling in B-cells. May be involved in BCR-induced apoptotic cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SH3 binding protein; SH3 domain-binding protein 5; SH3 domain-binding protein that preferentially associates with BTK; SH3-domain binding protein 5 (BTK-associated); SH3BP-5; Vascular endothelial cell-specific protein 18
Gene Aliases: AI606498; SAB; SH3BP-5; SH3BP5; Vesp18
UniProt ID: (Human) O60239, (Rat) Q91Y80, (Mouse) Q9Z131
Entrez Gene ID: (Human) 9467, (Bovine) 618056, (Rat) 117186, (Mouse) 24056
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support