Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EEN; EEN fusion partner of MLL; endophilin A2; Endophilin-2; Endophilin-A2; extra 11-19 leukemia fusion; Extra eleven-nineteen leukemia fusion gene protein; MGC111371; SH3 domain containing GRB2 like 1; SH3 domain protein 2B; SH3 domain-containing GRB2-like protein 1; SH3-containing Grb-2-like 1 protein; SH3-domain GRB2-like 1; SH3p8
Gene Aliases: CNSA1; EEN; SH3D2B; SH3GL1; SH3P8
UniProt ID: (Human) Q99961, (Rat) O35964
Entrez Gene ID: (Human) 6455, (Rat) 81922
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support