Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: SRAKLSMINT MSKIRGQEKG PGYPQAEALL AEAMLKFGRE LGDDCNFGPA LGEVGEAMRE L
Enables identical protein binding activity. Involved in negative regulation of blood-brain barrier permeability; negative regulation of gene expression; and negative regulation of protein phosphorylation. Located in perinuclear region of cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA335L15.1; bA335L15.1 (SH3-domain GRB2-like 2); EEN-B1; endophilin; Endophilin A1 BAR domain; Endophilin-1; Endophilin-A1; FLJ20276; FLJ25015; SH3 domain containing GRB2 like 2; SH3 domain protein 2A; SH3 domain-containing GRB2-like protein 2; SH3-domain GRB2-like 2; SH3p4
Gene Aliases: CNSA2; EEN-B1; SH3D2A; SH3GL2; SH3P4
UniProt ID: (Human) Q99962, (Rat) O35179
Entrez Gene ID: (Human) 6456, (Rat) 116743
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support