Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDFNMKKLAS DAGIFFTRAV QFTEEKFGQA EKTELDAHFE NLLARADSTK NWTEKILRQT EVLLQPNPSA RVEEFLYEKL DRKVPSRVTN GELLAQYMAD AASELGPTTP YGKTLIKVAE AEKQLGAAER DFIHTASISF LTPLRNFLEG DWKTISKERR LLQNRRLDLD ACKARLKKAK AAEAKATTVP DFQETRPRNY ILSASASALW NDEVDKAEQE LRVAQTEFDR QAEVTRLLLE GISSTHVNHL RCLHEFVKSQ TTYYAQCYRH MLDLQKQLGR FPGTFVGTTE PASPPLSSTS PTTAAATMPV VPSVASLAPP GEASLCLEEV APPASGTRKA RVLYDYEAAD SSELALLADE LITVYSLPGM DPDWLIGERG NKKGKVPVTY LELLS
SH3GLB2 (SH3 Domain Containing GRB2 Like, Endophilin B2) is a Protein Coding gene. Among its related pathways are Endocytosis. An important paralog of this gene is SH3GLB1. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endophilin-B2; retinoid receptor-induced gene 1; SH3 domain-containing GRB2-like protein B2; SH3-containing protein SH3GLB2; SH3-domain GRB2-like endophilin B2; sh3glb2 {ECO:0000312|EMBL:AAH87652.1}; stimulator of prostatic adenocarcinoma-specific T-cells-1
Gene Aliases: KIAA1848; PP578; PP6569; PP9455; RRIG1; SH3GLB2; SPAS-1
UniProt ID: (Human) Q9NR46, (Mouse) Q8R3V5
Entrez Gene ID: (Human) 56904, (Rabbit) 100345396, (Pig) 100156344, (Mouse) 227700, (Rat) 311848
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support