Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: IKLRPRSIEV ENDFLPVEKT IGKKLPATTA TPDSSKTEMD SRTKSKDYCK VIFPYEAQND DELTIKEGDI VTLINKDCID VGWWE
This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Cbl-interacting protein; Cbl-interacting protein of 85 kDa; CD2-binding protein 3; CD2BP3; HSB-1; Human Src family kinase-binding protein 1; migration-inducing gene 18; SH3 domain-containing kinase-binding protein 1; SH3-domain kinase binding protein 1; Src family kinase-binding protein 1; src-related kinase binding protein-1
Gene Aliases: CD2BP3; CIN85; GIG10; HSB-1; HSB1; MIG18; SH3KBP1
UniProt ID: (Human) Q96B97
Entrez Gene ID: (Human) 30011
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support