Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Adapter protein involved in invadopodia and podosome formation, extracellular matrix degradation and invasiveness of some cancer cells. Binds matrix metalloproteinases (ADAMs), NADPH oxidases (NOXs) and phosphoinositides. Acts as an organizer protein that allows NOX1- or NOX3-dependent reactive oxygen species (ROS) generation and ROS localization. In association with ADAM12, mediates the neurotoxic effect of amyloid-beta peptide.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adapter protein TKS5; adaptor protein TKS5; Five SH3 domain-containing protein; SH3 and PX domain-containing protein 2A; SH3 multiple domains 1; SH3 multiple domains protein 1; Tyrosine kinase substrate with five SH3 domains
Gene Aliases: FISH; KIAA0418; SH3MD1; SH3PXD2A; TKS5
UniProt ID: (Human) Q5TCZ1
Entrez Gene ID: (Human) 9644
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support