Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects a 225 kDa* protein corresponding to the molecular mass of Shank1 on SDS-PAGE immunoblots of adult mouse brain.
Seems to be an adapter protein in the postsynaptic density (PSD) of excitatory synapses that interconnects receptors of the postsynaptic membrane including NMDA-type and metabotropic glutamate receptors via complexes with GKAP/PSD-95 and Homer, respectively, and the actin-based cytoskeleton. May play a role in the structural and functional organization of the dendritic spine and synaptic junction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; OTTHUMP00000174437; SAPAP-interacting protein synamon; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains protein 1; SH3/ankyrin domain gene 1; SHANK 1; Shank1; Somatostatin receptor-interacting protein; SPANK 1; SPANK-1; SSTR-interacting protein; Synamon
Gene Aliases: SHANK1; Shank1a; SPANK-1; Spank1; SSTRIP; synamon
UniProt ID: (Human) Q9Y566, (Rat) Q9WV48, (Mouse) D3YZU1
Entrez Gene ID: (Human) 50944, (Rat) 78957, (Mouse) 243961
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support