Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Src homology 2 (SH2) domains bind specifically to tyrosine-phosphorylated proteins that temporally participate in signal transduction events. Shc-like protein (Sck) is a neuronal adaptor protein that contains an N-terminal PTB (phosphotyrosine binding) domain, a collagen homology (CH) domain, and a conserved C-terminal SH2 domain. Human Sck transcripts are present at high levels in liver, pancreas, prostate and ovary. In vascular endothelial cells, Sck participates in VEGF-induced signal transduction. Treatment of human umbilical vein endothelial (HUVEC) cells with VEGF induces recruitment of Sck to tyrosine-1175 of the kinase insert domain-containing receptor (KDR) and enhances Sck tyrosine phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neuronal Shc adaptor homolog; Protein Sck; Protein Sli; SH2 domain protein C2; SHC (Src homology 2 domain containing) transforming protein 2; SHC-transforming protein 2; SHC-transforming protein B; src homology 2 domain-containing transforming protein C2; Src homology 2 domain-containing-transforming protein C2
Gene Aliases: 6720466E06; SCK; SHC2; SHCB; SLI
UniProt ID: (Human) P98077, (Mouse) Q8BMC3, (Rat) O70142
Entrez Gene ID: (Human) 25759, (Mouse) 216148, (Rat) 314612
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support