Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SHISA9 Isoform X2 and its gene ontology information indicates that it is invovled in the cell membrane and may play an integral component of the membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2700045P11Rik; CKAMP44; cystine-knot AMPAR modulating protein; Cystine-knot AMPAR modulating protein of 44 kDa; FLJ10015; Protein shisa-9; shisa homolog 9; SHISA9
Gene Aliases: 2700045P11Rik; CKAMP44; SHISA9
UniProt ID: (Human) B4DS77, (Mouse) Q9CZN4
Entrez Gene ID: (Human) 729993, (Mouse) 72555
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support