Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: EKVLEACSIA CNKNTCPGDR SALRPSGLRL GTPALTSRGL LEKDFQKVAH FIHRGIELTL QIQSDTGVRA TLKEFKERLA GDKYQAAVQA LREEVESFAS LFPLPGLPD
This gene encodes the cellular form of serine hydroxymethyltransferase, a pyridoxal phosphate-containing enzyme that catalyzes the reversible conversion of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. This reaction provides one carbon units for synthesis of methionine, thymidylate, and purines in the cytoplasm. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing of this gene results in 2 transcript variants encoding 2 different isoforms. Additional transcript variants have been described, but their biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoplasmic serine hydroxymethyltransferase; Glycine hydroxymethyltransferase; MGC15229; MGC24556; serine hydroxymethyltransferase 1 (soluble); Serine hydroxymethyltransferase, cytosolic; Serine methylase; SHMT
Gene Aliases: CSHMT; SHMT; SHMT1
UniProt ID: (Human) P34896
Entrez Gene ID: (Human) 6470
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support